DL-Panthenol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
A149247-1000G
A149247-100G
A149247-25G
A149247-500G
76798-560EA
187
USD
76798-560
76798-562
76798-564
76798-566
DL-Panthenol 98%
DL-Panthenol
Precaución: for Research Use Only
Formula:
C₉H₁₉NO₄ MW: 205.25 g/mol |
MDL Number:
MFCD00002944 CAS Number: 16485-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...