M-Aminobenzyl cyanide 96%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Aminophenyl)acetonitrile

A147618-1G A147618-250MG
76798-510EA 77.45 USD
76798-510 76798-512
M-Aminobenzyl cyanide 96%
M-Aminobenzyl cyanide

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.163 g/mol
MDL Number: MFCD03265299
CAS Number: 4623-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR