N-Fmoc-N'-Cbz-D-Lysine 97%

Supplier: Ambeed
Warning

Synonyms: Nepsilon-Benzyloxycarbonyl-Nα-Fmoc-D-lysine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((benzyloxy)carbonyl)amino)hexanoic acid, Fmoc-D-Lys(Z)-OH

A147401-1G A147401-250MG A147401-25G A147401-5G
76798-484EA 10 USD
76798-484 76798-486 76798-488 76798-490
N-Fmoc-N'-Cbz-D-Lysine 97%
N-Fmoc-N'-Cbz-D-Lysine

Precaución: for Research Use Only

Formula: C₂₉H₃₀N₂O₆
MDL Number: MFCD00065663
CAS Number: 110990-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR