Ethyl-trans-4-bromocinnamate 98%

Supplier: Ambeed
Warning

Synonyms: (E)-Ethyl 3-(4-bromophenyl)acrylate

A146674-100G A146674-10G A146674-1G A146674-250MG A146674-25G A146674-5G
76798-440EA 580.32 USD
76798-440 76798-442 76798-444 76798-446 76798-448 76798-450
Ethyl-trans-4-bromocinnamate 98%
Ethyl-trans-4-bromocinnamate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₂
MW: 255.11 g/mol
MDL Number: MFCD00012226
CAS Number: 24393-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR