Methyl methyl-4-nitro-1H-imidazole-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A145349-100MG
A145349-1G
A145349-250MG
A145349-5G
76798-412EA
10
USD
76798-412
76798-414
76798-416
76798-418
Methyl methyl-4-nitro-1H-imidazole-2-carboxylate 98%
Methyl methyl-4-nitro-1H-imidazole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₇N₃O₄ |
MDL Number:
MFCD08275078 CAS Number: 169770-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...