4-Quinolinecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde

A143848-10G A143848-1G A143848-250MG A143848-25G A143848-5G
76798-388EA 71.09 USD
76798-388 76798-390 76798-392 76798-394 76798-396
4-Quinolinecarbaldehyde 98%
4-Quinolinecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.17 g/mol
MDL Number: MFCD00006781
CAS Number: 4363-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR