2-Thien-2-ylbenzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Thiophen-2-yl)benzaldehyde

A143393-1G A143393-250MG
76798-384EA 107.16 USD
76798-384 76798-386
2-Thien-2-ylbenzaldehyde 95%
2-Thien-2-ylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈OS
MW: 188.25 g/mol
MDL Number: MFCD04039148
CAS Number: 99902-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR