1-Acenaphthenol 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dihydroacenaphthylen-1-ol

A143000-1G A143000-25G
76798-380EA 35.01 USD
76798-380 76798-382
1-Acenaphthenol 98%
1-Acenaphthenol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
MDL Number: MFCD00003808
CAS Number: 6306-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR