(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-nitrophenyl)propanoic acid

A142355-100MG A142355-1G A142355-250MG
76798-368EA 46.68 USD
76798-368 76798-370 76798-372
(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid 95%
(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂O₆
MW: 432.43 g/mol
MDL Number: MFCD03428007
CAS Number: 507472-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR