H-Phe(2,5-DiCl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: L-2.5-Dichlorophenylalanine, (S)-2-Amino-3-(2,5-dichlorophenyl)propanoic acid

A142072-1G A142072-250MG A142072-25G A142072-5G
76798-360EA 89.12 USD
76798-360 76798-362 76798-364 76798-366
H-Phe(2,5-DiCl)-OH 95%
H-Phe(2,5-DiCl)-OH

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO₂
MW: 234.2 g/mol
MDL Number: MFCD07372017
CAS Number: 754971-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR