N-Fmoc-3-(4-thiazolyl)-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-3-(4-thiazolyl)-Alanine, Fmoc-β-(4-Thiazolyl)-Ala-OH, Fmoc-L-(4-thiazolyl)-Alanine

A142002-100MG A142002-250MG A142002-25G A142002-5G A142002-10G
76798-352EA 25 USD
76798-352 76798-354 76798-356 76798-358 77667-060
N-Fmoc-3-(4-thiazolyl)-L-alanine 97%
N-Fmoc-3-(4-thiazolyl)-L-alanine

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₂O₄S
MDL Number: MFCD00672568
CAS Number: 205528-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR