L-Aspartic acid monosodium salt monohydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium (S)-3-amino-3-carboxypropanoate hydrate, Sodium L-aspartate monohydrate, L-Aspartic acid sodium salt monohydrate
A141709-1000G
A141709-100G
A141709-500G
76798-324EA
28.65
USD
76798-324
76798-326
76798-328
L-Aspartic acid monosodium salt monohydrate 98%
L-Aspartic acid monosodium salt monohydrate
Precaución: for Research Use Only
Formula:
C₄H₆NNaO₄·H₂O MW: 173.11 g/mol (155.10 anhy) |
MDL Number:
MFCD00152960 CAS Number: 323194-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...