4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-4-Amc-OH

A141531-250MG A141531-1G A141531-5G
76798-320EA 41 USD
76798-320 77878-999 77879-000
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid 95%
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₄
MDL Number: MFCD01862347
CAS Number: 188715-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR