AMAS (N-Succinimidyl maleimidoacetate) 97%

Supplier: Ambeed
Warning

Synonyms: Maleimidoacetic acid N-hydroxysuccinimide ester, AMAS, N-(alpha-Maleimidoacetoxy)succinimide, Maleimidoacetic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetate

A141381-1G A141381-250MG A141381-5G A141381-100MG A141381-25G
76798-306EA 141.1 USD
76798-306 76798-308 76798-310 77600-054 77667-048
AMAS (N-Succinimidyl maleimidoacetate) 97%
AMAS (N-Succinimidyl maleimidoacetate)

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₆
MW: 252.18 g/mol
MDL Number: MFCD00133523
CAS Number: 55750-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR