Methyl 2-chloro-3-methylisonicotinate 97%

Supplier: Ambeed
Warning

A141296-100MG A141296-10G A141296-1G A141296-250MG A141296-25G A141296-5G
76798-294EA 13 USD
76798-294 76798-296 76798-298 76798-300 76798-302 76798-304
Methyl 2-chloro-3-methylisonicotinate 97%
Methyl 2-chloro-3-methylisonicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD08275065
CAS Number: 787596-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR