Methyl-2-(bromomethyl)-3-furoate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-(bromomethyl)furan-3-carboxylate

A141062-1G A141062-250MG A141062-5G
76798-280EA 102 USD
76798-280 76798-282 77878-996
Methyl-2-(bromomethyl)-3-furoate 97%
Methyl-2-(bromomethyl)-3-furoate

Precaución: for Research Use Only

Formula: C₇H₇BrO₃
MW: 219.04 g/mol
MDL Number: MFCD02180747
CAS Number: 53020-08-9
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR