D-Cysteine methyl ester hydrochloride 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
H-D-Cys-OMe·HCl, H-D-Cys-OMe.HCl, (S)-Cysteine methyl ester hydrochloride, Methyl D-cysteinate HCl, Methyl D-cysteinate hydrochloride
A141060-10G
A141060-1G
A141060-250MG
A141060-25G
A141060-5G
A141060-100G
76798-270EA
31
USD
76798-270
76798-272
76798-274
76798-276
76798-278
77667-036
D-Cysteine methyl ester hydrochloride 97%
D-Cysteine methyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉NO₂S MW: 171.64 g/mol |
MDL Number:
MFCD00672724 CAS Number: 70361-61-4 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...