Guanidine phosphate 99%

Supplier: Ambeed
Warning

A140923-100G A140923-500G
76798-256EA 6.37 USD
76798-256 76798-258
Guanidine phosphate 99%
Guanidine phosphate

Precaución: for Research Use Only

Formula: CH₈N₃O₄P
MW: 157.07 g/mol
MDL Number: MFCD00066148
CAS Number: 5423-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR