7-Hydroxy-6-methoxy-8-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one 98%

Supplier: Ambeed
Warning

A140824-10MG A140824-1MG A140824-25MG
76798-250EA 28.65 USD
76798-250 76798-252 76798-254
7-Hydroxy-6-methoxy-8-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one 98%
7-Hydroxy-6-methoxy-8-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₁₀
MW: 370.30 g/mol
MDL Number: MFCD00010093
CAS Number: 524-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR