L(-)-Menthol 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
A140621-100G
A140621-10G
A140621-25G
A140621-500G
A140621-1000G
76798-234EA
11
USD
76798-234
76798-236
76798-238
76798-240
77667-028
L(-)-Menthol 99+%
L(-)-Menthol
Precaución: for Research Use Only
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Boiling Pt: 212 Flash Pt: 96 |
MDL Number:
MFCD00062979 CAS Number: 2216-51-5 Merck Index: 13,05861 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...