(S)-(+)-Ethyl-3-hydroxybutyrate 95%

Supplier: Ambeed
Warning

Synonyms: (S)-Ethyl-3-hydroxybutyrate, Ethyl (S)-3-hydroxybutanoate, Ethyl (S)-(+)-3-hydroxybutyrate, (S)-Ethyl 3-hydroxybutanoate, Ethyl (S)-3-hydroxybutyrate

A140265-10G A140265-1G A140265-5G A140265-25G
76798-218EA 27 USD
76798-218 76798-220 76798-222 77566-820
(S)-(+)-Ethyl-3-hydroxybutyrate 95%
(S)-(+)-Ethyl-3-hydroxybutyrate

Precaución: For research use only.

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
MDL Number: MFCD00066206
CAS Number: 56816-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR