Imidazole-4-carboxamide 97%

Supplier: Ambeed
Warning

A140215-1G A140215-250MG A140215-5G A140215-100MG
76798-206EA 25 USD
76798-206 76798-208 76798-210 77600-036
Imidazole-4-carboxamide 97%
Imidazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MDL Number: MFCD04113065
CAS Number: 26832-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR