Hexamethonium bromide 99%

Supplier: Ambeed
Warning

A139736-25G A139736-50MG A139736-10MG A139736-500G A139736-100G
76798-178EA 11 USD
76798-178 76798-180 77600-028 77667-020 77878-986
Hexamethonium bromide 99%
Hexamethonium bromide

Precaución: for Research Use Only

Formula: C₁₂H₃₀Br₂N₂
MDL Number: MFCD00011787
CAS Number: 55-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR