1-(4-Methoxyphenyl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid

A139043-10G A139043-1G A139043-250MG A139043-25G A139043-5G
76798-158EA 79 USD
76798-158 76798-160 76798-162 76798-164 76798-166
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid 97%
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD00019223
CAS Number: 16728-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR