4-(1H-Pyrazol-1-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

A138603-100G A138603-10G A138603-1G A138603-250MG A138603-25G A138603-5G
76798-140EA 292 USD
76798-140 76798-142 76798-144 76798-146 76798-148 76798-150
4-(1H-Pyrazol-1-yl)benzaldehyde 97%
4-(1H-Pyrazol-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MDL Number: MFCD02681938
CAS Number: 99662-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR