4-(2-Furyl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Furan-2-yl)aniline

A137810-100MG A137810-1G A137810-250MG A137810-5G
76798-118EA 65.78 USD
76798-118 76798-120 76798-122 77600-006
4-(2-Furyl)aniline 95%
4-(2-Furyl)aniline

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD04039072
CAS Number: 59147-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR