N-(1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)isobutyramide 95%
Supplier: Ambeed
|
Warning
|
A137389-10G
A137389-1G
A137389-25G
A137389-5G
76798-110EA
54
USD
76798-110
76798-112
76798-114
76798-116
N-(1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)isobutyramide 95%
N-(1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)isobutyramide
Precaución: for Research Use Only
Formula:
C₁₃H₁₉N₃O₅ |
MDL Number:
MFCD00079383 CAS Number: 110522-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...