4-(p-Tolyl)butyric acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(p-Tolyl)butanoic acid
A136609-100G
A136609-10G
A136609-1G
A136609-250MG
A136609-25G
A136609-5G
76798-090EA
288
USD
76798-090
76798-092
76798-094
76798-096
76798-098
76798-100
4-(p-Tolyl)butyric acid 97%
4-(p-Tolyl)butyric acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₂ MW: 178.23 g/mol |
MDL Number:
MFCD00021786 CAS Number: 4521-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...