Ethyl 2-phenyl-1H-indole-3-carboxylate 96%

Supplier: Ambeed
Warning

A136489-100MG A136489-1G A136489-250MG
76798-084EA 80.63 USD
76798-084 76798-086 76798-088
Ethyl 2-phenyl-1H-indole-3-carboxylate 96%
Ethyl 2-phenyl-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₂
MW: 265.30 g/mol
MDL Number: MFCD04038610
CAS Number: 36779-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR