DL-Lysine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Lys-OH.2HCl, 2,6-Diaminohexanoic acid dihydrochloride, DL-Lysine 2HCl

A136472-1G A136472-25G A136472-5G
76798-078EA 9.55 USD
76798-078 76798-080 76798-082
DL-Lysine dihydrochloride 98%
DL-Lysine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂O₂
MW: 219.11 g/mol
MDL Number: MFCD00037587
CAS Number: 617-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR