Dodecylamine 98%

Supplier: Ambeed
Danger

A136304-25G A136304-100G
76798-068EA 7 USD
76798-068 77878-970
Dodecylamine 98%
Dodecylamine

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₀CH₂NH₂
MDL Number: MFCD00008154
CAS Number: 124-22-1
UN: 3259
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR