L-Aspartic acid dibenzyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Asp(Obzl)-OBzl.HCl, H-Asp(OBzl)-OBzl·HCl

A136200-10G A136200-25G A136200-5G A136200-100G
76798-056EA 10 USD
76798-056 76798-058 76798-060 77878-969
L-Aspartic acid dibenzyl ester hydrochloride 97%
L-Aspartic acid dibenzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₄·HCl
MDL Number: MFCD01075101
CAS Number: 6327-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR