Methyl 2,4-dimethylbenzoate 95%

Supplier: Ambeed
Warning

A136167-100G A136167-10G A136167-1G A136167-25G
76798-048EA 228.1 USD
76798-048 76798-050 76798-052 76798-054
Methyl 2,4-dimethylbenzoate 95%
Methyl 2,4-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
MDL Number: MFCD06203654
CAS Number: 23617-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR