Ethyl 2-amino-5-methylthiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A134830-1G A134830-250MG
76798-010EA 35.01 USD
76798-010 76798-012
Ethyl 2-amino-5-methylthiazole-4-carboxylate 97%
Ethyl 2-amino-5-methylthiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD01536173
CAS Number: 72054-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR