Gomisin A 99+%

Supplier: Ambeed
Warning

A134812-10MG A134812-1MG A134812-5MG A134812-100MG A134812-50MG
76798-004EA 96 USD
76798-004 76798-006 76798-008 77878-958 77878-959
Gomisin A 99+%
Gomisin A

Precaución: for Research Use Only

Formula: C₂₃H₂₈O₇
MDL Number: MFCD01941645
CAS Number: 58546-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR