Methyl 2-bromo-6-chlorobenzoate 95%

Supplier: Ambeed
Warning

A134722-1G A134722-250MG A134722-5G A134722-25G
76797-996EA 30 USD
76797-996 76797-998 77566-644 77878-957
Methyl 2-bromo-6-chlorobenzoate 95%
Methyl 2-bromo-6-chlorobenzoate

Precaución: For research use only.

Formula: C₈H₆BrClO₂
MDL Number: MFCD03789160
CAS Number: 685892-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR