4'-Methoxyacetoacetanilide 95%

Supplier: Ambeed
Warning

A134468-10G A134468-25G
76797-992EA 6.37 USD
76797-992 76797-994
4'-Methoxyacetoacetanilide 95%
4'-Methoxyacetoacetanilide

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.2300 g/mol
MDL Number: MFCD00008783
CAS Number: 5437-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR