Ethyl 2-(3,5-dimethoxyphenyl)acetate 98%

Supplier: Ambeed
Warning

A134132-100MG A134132-10G A134132-1G A134132-250MG
76797-982EA 15.92 USD
76797-982 76797-984 76797-986 76797-988
Ethyl 2-(3,5-dimethoxyphenyl)acetate 98%
Ethyl 2-(3,5-dimethoxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.25 g/mol
MDL Number: MFCD11111879
CAS Number: 65976-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR