6-Methoxy-2-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxynaphthalen-2-ol, Naproxen Impurity H

A133766-100MG A133766-1G A133766-250MG A133766-5G A133766-25G
76797-962EA 20.16 USD
76797-962 76797-964 76797-966 76797-968 77599-920
6-Methoxy-2-naphthol 98%
6-Methoxy-2-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MW: 174.2000 g/mol
MDL Number: MFCD00088677
CAS Number: 5111-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR