(-)-Menthyl acetate 98%

Supplier: Ambeed
Warning

Synonyms: L-Menthyl acetate, L-Menthol acetate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate, (R)-(-)-Menthyl acetate, (-)-Acetoxy-p-menthane, (5R)-2-Isopropyl-5-methylcyclohexyl acetate, (1R)-(−)-Menthyl acetate

A133176-500G A133176-25G
76797-956EA 101 USD
76797-956 77666-912
(-)-Menthyl acetate 98%
(-)-Menthyl acetate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00001482
CAS Number: 2623-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR