2-Hydroxyquinoline-4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxycinchoninic Acid
A132791-10G
A132791-1G
A132791-25G
A132791-5G
A132791-100G
76797-940EA
21.22
USD
76797-940
76797-942
76797-944
76797-946
77624-120
2-Hydroxyquinoline-4-carboxylic acid 97%
2-Hydroxyquinoline-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₇NO₃ MW: 189.17 g/mol |
MDL Number:
MFCD00023942 CAS Number: 15733-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...