Isoxazol-5-ylamine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Aminoisoxazole, Isoxazol-5-amine

A131796-100MG A131796-1G A131796-250MG A131796-5G
76797-926EA 33.95 USD
76797-926 76797-928 76797-930 76797-932
Isoxazol-5-ylamine 98%
Isoxazol-5-ylamine

Precaución: for Research Use Only

Formula: C₃H₄N₂O
MW: 84.08 g/mol
MDL Number: MFCD00128192
CAS Number: 14678-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR