(2R)-N-[3-[2-[2-[3-[[(2R)-2,4-Dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulfanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamide 95% (by HPLC) (contains < 20% H₂O)

Supplier: Ambeed
Danger

A131343-100G A131343-25G A131343-1G
76797-888EA 489 USD
76797-888 76797-890 77624-114
(2R)-N-[3-[2-[2-[3-[[(2R)-2,4-Dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulfanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamide 95% (by HPLC) (contains < 20% H₂O)
(2R)-N-[3-[2-[2-[3-[[(2R)-2,4-Dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulfanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamide

Precaución: for Research Use Only

Formula: C₂₂H₄₂N₄O₈S₂
MDL Number: MFCD07357234
CAS Number: 16816-67-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR