4-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

A131265-250MG A131265-5G A131265-1G
76797-884EA 23 USD
76797-884 76797-886 77878-946
4-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 97%
4-(2-Methyl-1H-imidazol-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MDL Number: MFCD05864665
CAS Number: 88427-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR