Imipramine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine hydrochloride

A131108-1MG A131108-25MG A131108-1G
76797-874EA 5.31 USD
76797-874 76797-876 77666-882
Imipramine hydrochloride 95%
Imipramine hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₄N₂·HCl
MW: 316.87 g/mol
MDL Number: MFCD00012669
CAS Number: 113-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR