DL-Carnitine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride

A130298-10MG A130298-50MG
76797-848EA 5.31 USD
76797-848 76797-850
DL-Carnitine hydrochloride 97%
DL-Carnitine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₃
MW: 197.66
MDL Number: MFCD00011904
CAS Number: 461-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR