3-Fluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-(pyrolidine-1-carbonyl)phenylboronic acid, (3-Fluoro-4-(pyrrolidine-1-carbonyl)phenyl)boronic acid

A129821-1G A129821-250MG A129821-5G
76797-826EA 106.09 USD
76797-826 76797-828 76797-830
3-Fluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%
3-Fluoro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BFNO₃
MW: 237.0400 g/mol
MDL Number: MFCD08436024
CAS Number: 874289-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR