Nitrofurazone 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Nitrofurfural Semicarbazone, 2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide, 5-Nitro-2-Furaldehyde Semicarbazone
A129741-1G
A129741-1MG
A129741-5G
A129741-100G
76797-820EA
9.55
USD
76797-820
76797-822
76797-824
77666-852
Nitrofurazone 98+%
Nitrofurazone
Precaución: for Research Use Only
Formula:
C₆H₆N₄O₄ MW: 198.14 g/mol |
MDL Number:
MFCD00003225 CAS Number: 59-87-0 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...