H-P-Fluoro-DL-Phe-OMe·HCl 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-3-(4-fluorophenyl)propanoate hydrochloride, H-p-Fluoro-DL-Phe-OMe·HCl

A129634-100G A129634-10G A129634-1G A129634-250MG A129634-25G A129634-5G
76797-806EA 1002.56 USD
76797-806 76797-808 76797-810 76797-812 76797-814 76797-816
H-P-Fluoro-DL-Phe-OMe·HCl 97%
H-P-Fluoro-DL-Phe-OMe·HCl

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClFNO₂
MW: 233.667 g/mol
MDL Number: MFCD00182718
CAS Number: 64282-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR