6-Fluoro-4-hydroxyquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A129189-1G A129189-250MG
76797-798EA 6.37 USD
76797-798 76797-800
6-Fluoro-4-hydroxyquinoline-3-carboxylic acid 97%
6-Fluoro-4-hydroxyquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆FNO₃
MW: 207.1600 g/mol
MDL Number: MFCD00973836
CAS Number: 343-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR